missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cucurbitacin E, MedChemExpress
Description
Cucurbitacin E is a natural compound which from the climbing stem of Cucumic melo L. Cucurbitacin E significantly suppresses the activity of the cyclin B1/CDC2 complex.
Specifications
Specifications
| Chemical Name or Material | Cucurbitacin E |
| CAS | 18444-66-1 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C32H44O8 |
| Quantity | 5 mg |
| Synonym | α-Elaterin α-Elaterine |
| Solubility Information | DMSO : 50 mg/mL (89.82 mM; Need ultrasonic) |
| SMILES | O[C@H](C1)[C@@]([C@@](C)(O)C(/C=C/C(OC(C)=O)(C)C)=O)([H])[C@](C2)(C)[C@]1(C)[C@]3([H])CC=C4C(C)(C)C(C(O)=C[C@@]4([H])[C@]3(C)C2=O)=O |
| Molecular Weight (g/mol) | 556.69 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?