Learn More
Sartorius Cubis™ II Micro Balance, MCE User Interface
BIOPHARMA
Product Code. 16437185 Shop All Sartorius Products
Change view
Click to view available options
Capacity:
1.1 g, 2.1 g, 3.1 g
10.1 g
Model:
Draft shield F: Filtered
Draft shield M: Motorized
Includes:
Balance, Accessories, Power supply, Manual, Factory test report
Balance, Accessories, Power supply, Manual, Factory test report, QP99 software
Unit Size:
1 unit
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Model Includes Unit Size
16437185 10.1 g Draft shield M: Motorized Balance, Accessories, Power supply, Manual, Factory test report Each
16427185 10.1 g Draft shield F: Filtered Balance, Accessories, Power supply, Manual, Factory test report Each
16407315 1.1 g, 2.1 g, 3.1 g Draft shield F: Filtered Balance, Accessories, Power supply, Manual, Factory test report Each
16283262 1.1 g, 2.1 g, 3.1 g Draft shield M: Motorized Balance, Accessories, Power supply, Manual, Factory test report, QP99 software 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16437185 Supplier Sartorius Supplier No. MCE10.6S2S00M
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
Estimated Shipment: 14-01-2027
Cubis™ II Micro Balance features fully customizable hardware, software, and connectivity
  • Monolithical weighing system
  • Integrated climate sensors
  • Individual sample holders
  • Error-free operation
  • Individual QApp workflows
  • Motorized auto-leveling
  • Full end-to-end data integrity
  • Integrated audit-trail
  • State-of-the-art user management
  • Excellent service support
  • Integrated status center
  • Integrated service functions
  • Preventive maintenance on accredited standards
  • TFT touch screen operation with readable display and easy to operate user interface
  • Software Factory installed basic set of essential weighing applications
  • Operation Activated by touch key, touch-free using IR sensor (draft shield M), or gesture sensor (optional) learning capability
Warranty
10-year warranty
TRUSTED_SUSTAINABILITY
Specifications
Connection Type 2 USB A ports, 1 USB B port, 1USP C port, 1-9 pin RS 232 connection, 1 ethernet port
Capacity 10.1 g
Display Type Color TFT Touchscreen
Readability 1 μg
Includes Balance, Accessories, Power supply, Manual, Factory test report
Linearity ±3 μg
Material Die-cast Aluminum, Plastic PBT, Optiwhite Float Glass and Stainless Steel, Aluminum Trim
Model Draft shield M: Motorized
Repeatability ±5 μg
Stabilization Time 5 sec.
Type Micro Balance
Warranty 10 years
Length (Metric) 340/355 mm weighinh/electrical module
Width (Metric) 139/240 mm
Height (Metric) 129/61 mm weighing/electrical module
Weight (Metric) 7.2, 7.9 kg
Electrical Requirements 120/240 V
Voltage 100/240 V ±10%
Frequency 50/60 Hz
Legal for Trade Class I