Learn More
Tocris Bioscience™ CTPI 2
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
CTPI 2 is a selective mitochondrial citrate transporter (SLC25A1) inhibitor with a KD of 3.5 μM. CTPI 2 inhibits the proliferation of H1299 cells in a SLC25A1-dependent manner. It also re-sensitizes T1, T2, and T4 cells to Cisplatin (Cat. No. 2251). CTPI 2 inhibits tumor growth in in vivo models of non-small cell lung cancer (NSCLC). In high-fat diet fed mice CTPI 2 regulates glycolysis, prevents steatohepatitis and normalizes glucose tolerance and insulin sensitivity. CTPI 2 influences the inflammatory pathway by inhibiting IL-6 and TNFα production and M1 macrophage polarization.
Specifications
Specifications
| Chemical Name or Material | 2-[[(4-Chloro-3-nitrophenyl)sulfonyl]amino]benzoic acid |
| CAS | 68003-38-3 |
| Quantity | 50 mg |
| Target | Other Ion Pump/Transporter Inhibitors |
| Molecular Formula | C13H9ClN2O6S |
| Purity | 98% |