Learn More
Multiroir Controlec™ Crystal Multiroir Modular Drawer
Product Code. 30486999
Change view
Click to view available options
Capacity (Metric):
5 L
Unit Size:
Pack of 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
30486999 5 L Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30486999 Supplier Multiroir Controlec™ Supplier No. 11108MU05000C
Estimated Shipment: 04-01-2027
Perfectly modular and combinable. Multiroir Controlec™ Crystal Multiroir Modular Drawer combines exceptional transparency, high impact resistance and the possibility for autoclave sterilization.
  • Perfectly modular and stackable—combinable on all four sides
  • Interchangeable drawers and sleeves
  • Made of high transparency polycarbonate for easy viewing
  • Strong and sturdy with high resistance to shocks and UV
  • Can be sterilized in an autoclave for use in sterile/cleanroom environments
  • Integrated under-grip handle made of the plastic material
  • Large single drawer and sleeve with capacity of 5 liters
  • Identical size sleeve with 2 small drawers also available
Specifications
Capacity (Metric) 5 L
Certifications/Compliance Compliance with European standards 
Closure Color Transparent
Closure Type Under-Grip Handle
Color Transparent
Material Polycarbonate
Shape Rectangular
Includes Drawer and sleeve
No. of Compartments 1
Stackable Yes
Length (Metric) Exterior 355 mm
Length (Metric) Interior 320 mm
Width (Metric) Exterior 185 mm
Width (Metric) Interior 172 mm
Height (Metric) Exterior 101 mm
Height (Metric) Interior 92 mm