Learn More
Thermo Scientific™ Cryoware Labels
Product Code. 10634161
Change view
Click to view available options
Dimensions (L x W):
25 x 50 mm
Unit Size:
Pack of 200
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
10634161 25 x 50 mm Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10634161 Supplier Thermo Scientific™ Supplier No. 50400002
  • ONLINE EXCLUSIVE PRICE
    67.80 EUR
    59.50 EUR / Pack of 200
    Save 8.30 EUR
    12% Off
In Stock
Easily and permanently identify samples and storage boxes with Thermo Scientific™ Cryoware Labels.
  • 25 x 50mm cloth labels
  • Accept markers or ballpoint pen
  • Specifically designed for use at ultra-low temperatures
  • Adhere to plastic and cardboard without shrinking or peeling
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Not Autoclavable
Adhesive Type Cold-temperature
Color White
Material Nylon Cloth
Sterility Non-sterile
Latex Free Yes
Dimensions (L x W) 25 x 50 mm
Voltage 0/1 V