Learn More
Epredia™ CryoStar™ NX70 Specimen Chucks
Product Code. 12698213 Shop All Epredia Products
Change view
Click to view available options
Product Type:
30mm Specimen Chuck, Blue
30mm Specimen Chuck, Gold
30mm Specimen Chuck, Green
30mm Specimen Chuck, Red
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
12698213 30mm Specimen Chuck, Blue Each
12687576 30mm Specimen Chuck, Red Each
12697576 30mm Specimen Chuck, Green Each
12607586 30mm Specimen Chuck, Gold Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 12698213 Supplier Epredia™ Supplier No. 715890
  • ONLINE EXCLUSIVE PRICE
    26.70 EUR
    24.79 EUR / Pack of 1
    Save 1.91 EUR
    7% Off
In Stock
Choose between multiple available colors for Epredia™; CryoStar™ Cryostat Specimen Chucks.
TRUSTED_SUSTAINABILITY
Specifications
Product Type 30mm Specimen Chuck, Blue
For Use With (Equipment) CryoStar NX50 and NX70