Learn More
Thermo Scientific™ CryoBox™ Freezer Racks
Product Code. 10326442
Change view
Click to view available options
Holds:
12 CryoBoxes/Rack
16 CryoBoxes/Rack
4 CryoBoxes/Rack
7 CryoBoxes/Rack
9 CryoBoxes/Rack
Design:
Horizontal
Vertical
Dimensions (L x W x H):
14.3 x 14 x 40.6 cm
14.3 x 14 x 50.2 cm
14.3 x 14 x 70.5 cm
14.3 x 67 x 14 cm
14.3. x 14 x 22.5 cm
22.5 x 44.4 x 14.3 cm
22.5 x 59.0 x 14.3 cm
8.4 x 8.3 x 22.5 cm
8.4 x 8.3 x 50.2 cm
Förpackningsstorlek:
Each
Pack of 4
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Holds Design Dimensions (L x W x H) Förpackningsstorlek
10326442 12 CryoBoxes/Rack Vertical 14.3 x 67 x 14 cm Each
10419272 4 CryoBoxes/Rack Vertical 14.3. x 14 x 22.5 cm Each
11877113 9 CryoBoxes/Rack Vertical 14.3 x 14 x 50.2 cm Pack of 4
10547571 12 CryoBoxes/Rack Horizontal 22.5 x 44.4 x 14.3 cm Each
10653791 16 CryoBoxes/Rack Horizontal 22.5 x 59.0 x 14.3 cm Each
10674321 4 CryoBoxes/Rack Vertical 8.4 x 8.3 x 22.5 cm Each
10419082 9 CryoBoxes/Rack Vertical 8.4 x 8.3 x 50.2 cm Each
10504831 4 CryoBoxes/Rack Vertical 14.3 x 14 x 40.6 cm Each
10718861 7 CryoBoxes/Rack Vertical 14.3 x 14 x 70.5 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 10326442 Leverantör Thermo Scientific™ Leverantörsnummer 367000
  • ONLINE EXCLUSIVE PRICE
    494.00 EUR
    435.00 EUR / Pack of 1
    Save 59.00 EUR
    11% Off
 Restricted Item
In Stock
Lägg i varukorgen
Lägg i varukorgen
Denna artikel kan inte returneras. Se returpolicy
Stack and organize standard cryogenic vial storage boxes with these freezer racks.
Stack and organize standard cryogenic vial storage boxes with these freezer racks. Thermo Scientific™ CryoBox™ Freezer Racks include durable Thermo Scientific Nalgene™ Stainless Steel CryoBox Racks and Thermo Scientific Nunc™ aluminum racks that fit most common upright and chest freezers.
Warranty: 90 days
TRUSTED_SUSTAINABILITY
Specifikationer
Color Silver
Dimensions (L x W x H) 14.3 x 67 x 14 cm
Holds 12 CryoBoxes/Rack
Design Vertical
Material Stainless Steel
Product Type Cryogenic Box Storage Rack, Vertical
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.