Learn More
Thermo Scientific™ Cryo Marker Pen Set
Product Code. 15611105 Shop All Thermo Scientific Products
Change view
Click to view available options
Color:
Black
Multi-color
Unit Size:
Pack of 4
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
15611105 Black Pack of 4
15289134 Multi-color Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15611105 Supplier Thermo Scientific™ Supplier No. 4000222
  • ONLINE EXCLUSIVE PRICE
    177.41 EUR
    159.00 EUR / Pack of 4
    Save 18.41 EUR
    10% Off
 Restricted Item
In Stock
Label cryogenic vials to -200°C (-328°F) with the Thermo Scientific™ Cryo Marker Pen Set. The ink is fast-drying, nonsmearing and permanent.
Label cryogenic vials to -200°C (-328°F) with the Thermo Scientific™ Cryo Marker Pen Set. The ink is fast-drying, nonsmearing and permanent.
  • Set includes 4 black pens.
TRUSTED_SUSTAINABILITY
Specifications
Type Cryo Marker Pen Set
Temperature Range -200°C, -328°F
Color Black
For Use With (Equipment) cryovials and boxes
Includes 4 black pens