Learn More
Fisherbrand™ PTFE Magnetic Stir Bar, Cross-shaped
Product Code. 11898932 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Dimensions (L x W x H):
10 x 10 x 5mm
20 x 20 x 8mm
25 x 25 x 9mm
30 x 30 x 10mm
38 x 38 x 11mm
Unit Size:
Pack of 10
Pack of 5
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) Unit Size
11898932 10 x 10 x 5mm Pack of 10
11521983 10 x 10 x 5mm Pack of 5
11808942 20 x 20 x 8mm Pack of 5
16255791 25 x 25 x 9mm Pack of 10
11828942 30 x 30 x 10mm Pack of 5
16275791 38 x 38 x 11mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 11898932
Estimated Shipment: 17-12-2026
Use with containers, vessel, stirrers, and stirring hot plates. The PTFE-coated magnet stirrers are stable for many general purpose applications. The bars are cross-shaped for improved stirring action.
  • Stable, general-purpose magnetic stirrer
  • Provides a deep, quiet vortex even at relatively high speeds
  • Autoclavable
Specifications
Autoclavable Autoclavable
Certifications/Compliance Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
Color White
Description PTFE encapsulated Alnico V magnet core
Dimensions (L x W x H) 10 x 10 x 5mm
For Use With (Equipment) Containers & Vessels / Stirrer Hot Plates
Height (Metric) 5mm
Length (English) 0.375 in.
Width (English) 0.375 in.
Width (Metric) 10mm
Coating PTFE
Length (Metric) 10 mm
Quantity 5/Pk.
Shape Cross
Type PTFE Stirrer Bars Cross