Learn More
Croconic acid, 98%
Product Code. p-7026037
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11424837 1 g 1g
11434837 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11424837 Supplier Thermo Scientific Alfa Aesar Supplier No. B21809.03
Add to basket
CAS: 488-86-8 | C5H2O5 | 142.066 g/mol
Croconic acid is used to prepare 1,3-bis-(2-dimethylamino-5-thienyl)croconine by reacting with dimethyl-thiophen-2-yl-amine. It is also used in organic synthesis and employed as a dye for biological research purposes. Further, it is involved in the preparation of ethers such as dimethyl croconate. In addition to this, it acts as a ligand and form coordination complexes with metals such as barium, copper, silver and lead.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Croconic acid is used to prepare 1,3-bis-(2-dimethylamino-5-thienyl)croconine by reacting with dimethyl-thiophen-2-yl-amine. It is also used in organic synthesis and employed as a dye for biological research purposes. Further, it is involved in the preparation of ethers such as dimethyl croconate. In addition to this, it acts as a ligand and form coordination complexes with metals such as barium, copper, silver and lead.
Solubility
Soluble in water and ethanol.
Notes
Light sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 488-86-8
Molecular Formula C5H2O5
Molecular Weight (g/mol) 142.066
MDL Number MFCD00181389
InChI Key RBSLJAJQOVYTRQ-UHFFFAOYSA-N
Synonym croconic acid, crocic acid, 4,5-dihydroxy-4-cyclopentene-1,2,3-trione, unii-6hgy35uxaa, 6hgy35uxaa, 4-cyclopentene-1,2,3-trione, 4,5-dihydroxy, croconicacid, 4,5-dihydroxy-4-cyclopentene-1,2,3-trione #
PubChem CID 546874
IUPAC Name 4,5-dihydroxycyclopent-4-ene-1,2,3-trione
SMILES C1(=C(C(=O)C(=O)C1=O)O)O
Specifications
Melting Point ∼280°C (decomposition)
Odor Odorless
Quantity 1 g
Solubility Information Soluble in water and ethanol.
Formula Weight 142.07
Percent Purity 98%
Chemical Name or Material Croconic acid