missing translation for 'onlineSavingsMsg'
Learn More

Crocetin, free acid, >90%, MP Biomedicals™

Product Code. 11434872 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
25 mg
Unit Size:
1mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11434872 1 mg 1mg
11444872 5 mg 5mg
11454872 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11434872 Supplier MP Biomedicals Supplier No. 0219354301
  • ONLINE EXCLUSIVE PRICE
    62.85 EUR
    59.85 EUR / 1mg
    Save 3.00 EUR
    4% Off
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 27876-94-4
Molecular Formula C20H24O4
Molecular Weight (g/mol) 328.408
InChI Key PANKHBYNKQNAHN-MQQNZMFNSA-N
Synonym crocetin, trans-crocetin, 8,8'-diapocarotenedioic acid, natural yellow 6, 8,8'-diapocarotene-8,8'-dioic acid, 8,8'-diapo-psi,psi-carotenedioic acid, ccris 7484, unii-20tc155l9c, 2e,4e,6e,8e,10e,12e,14e-2,6,11,15-tetramethylhexadeca-2,4,6,8,10,12,14-heptaenedioic acid, .alpha.-crocetin
PubChem CID 5281232
ChEBI CHEBI:3918
IUPAC Name (2E,4E,6E,8E,10E,12E,14E)-2,6,11,15-tetramethylhexadeca-2,4,6,8,10,12,14-heptaenedioic acid
SMILES CC(=CC=CC=C(C)C=CC=C(C)C(=O)O)C=CC=C(C)C(=O)O

Specifications

Color Orange
Assay Percent Range >90%
Quantity 1 mg
Formula Weight 328.4
Physical Form Powder
Chemical Name or Material Crocetin, free acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.