Learn More
Slice™ Craft Blades
Product Code. 16443619
Change view
Click to view available options
Product Type:
Curved Edge, Rounded Tip
Straight Edge, Pointed Tip
Straight Edge, Rounded Tip
Unit Size:
Pack of 4
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
16443619 Straight Edge, Pointed Tip Pack of 4
16433619 Straight Edge, Rounded Tip Pack of 4
16453619 Curved Edge, Rounded Tip Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16443619 Supplier Slice™ Supplier No. 10519
  • ONLINE EXCLUSIVE PRICE
    34.72 EUR
    29.44 EUR / Pack of 4
    Save 5.28 EUR
    15% Off
Estimated Shipment: 04-12-2026
Finger-friendly™ edge cuts materials effectively but is safer to touch than traditional blades
  • 4 single-edge blades per pack
  • Compatible w/ Slice™ craft handles, scalpel, Precision Knife
  • Compatible tool: Slice™ 10548
  • Finger-friendly™ edge stays sharp 11x longer
  • Reduces injuries, lowers costs
  • Non-sparking, non-conductive, non-magnetic
  • Safe up to 1600 degrees Celsius
  • Chemically inert, never rusts
  • No-tool blade change
  • Fewer blade changes = fewer injuries
Specifications
Product Type Straight Edge, Pointed Tip
For Use With (Equipment) Slice™ Handles: 10548, 10589, 10580, 10568
Length (English) 1.29 in.
Length (Metric) 32.8 mm
Material Zirconium Oxide
Color White
For Use With (Application) Safety Cutting
Includes 4 single-edge blades per pack
No. per Pack 4 Pk.
Weight (English) 0.02 oz.
Weight (Metric) 0.65 g
Width (English) 0.24 in.
Width (Metric) 6.15 mm