Learn More
Control Company™ Cradle and USB Cable for Logger-Trac™ Temp Mini Datalogger
Shop All Fisherbrand Traceable ProductsDescription
- Required accessory for the Logger-Trac™ Mini Datalogger.
- Cradle is constructed of plastic
- Includes USB cable for downloading data onto datalogger
Specifications
Specifications
| Includes | USB cable |
| Material | Plastic |
| For Use With (Equipment) | Logger-Trac™ Temp Mini Datalogger |