Learn More
Pal International™ CPE Fabric Overshoes
Product Code. 30158549
Change view
Click to view available options
Color:
Blue
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
30158549 Blue Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30158549 Supplier Pal International™ Supplier No. R21233AX
  • ONLINE EXCLUSIVE PRICE
    4.30 EUR
    4.15 EUR / Pack of 100
    Save 0.15 EUR
    3% Off
Estimated Shipment: 15-12-2026
Made from high quality embossed polyethylene providing strength and grip on slippery floors. Pal International™ CPE Overshoes are elasticised at the ankle ensuring a good fit and offering protection for floors and footwear in a range of industries.
  • Constructed of high-quality embossed polyethylene providing strength and grip
  • Large 35.6cm (14 in.) size and elasticized ankle ensure a good fit on most feet
  • Suitable for protecting floors and footwear and ensuring environmental hygiene in a range of industries
  • Come in 3 colors (red, white or blue) to support color-coded hygiene workwear
  • High-volume packs dispense 2,000 pairs
Specifications
Color Blue
For Use With (Application) Protecting floors and footwear
Length (English) 14 in.
Length (Metric) 35.56 cm
Material Chlorinated Polyethylene
Size Universal
Sole Features Smooth sole
Style CPE
Product Type CPE Overshoes