missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ CP7 Control Synthetic Peptide

Product Code. 18439771 Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
This item is not returnable. View return policy

Product Code. 18439771

Brand: Novus Biologicals™ NBP231231

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Contains gene inserts that are fully sequenced and produced with high quality

TRUSTED_SUSTAINABILITY

Specifications

Species Human
Purity >95%, by HPLC.
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Quantity 1 mg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.