Learn More
Thermo Scientific Chemicals Cover Slips, KBr, used with 19mm Aperture IR Cards, No. 43935
Brand: Thermo Scientific Chemicals 044527.KM
142.00 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Additional Details : Weight : 0.01000kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Cover Slips, KBr | |
25 Pk. |
RUO – Research Use Only