Learn More
Karl Hecht™ Assistent™ Cover Glasses for Hemocytometers
Product Code. 10647293
Change view
Click to view available options
Thickness (Metric):
0.4 mm
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Thickness (Metric) Unit Size
10647293 0.4 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10647293 Supplier Karl Hecht™ Supplier No. 40415710
Estimated Shipment: 04-12-2026
Prevents color distortion due to super-transparent and colorless construction. Karl Hecht™ Assistent™ Cover Glasses for Hemocytometers have a refractive index.
  • Excellent flatness
  • High spectral transmission
  • Optimum resistance to chemical attack
  • Low levels of inclusions, striae, bubbles, streaks, etc.
Specifications
Product Type Cover Glass
Thickness (Metric) 0.4 mm
For Use With (Equipment) For Hemacytometer