Learn More
Hirschmann™ Counting Chambers with Clamps
Product Code. 10345511
Change view
Click to view available options
Product Type:
Counting Chamber (Burker)
Counting Chamber (Neubauer)
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
10345511 Counting Chamber (Burker) Each
10049630 Counting Chamber (Neubauer) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10345511 Supplier Hirschmann™ Supplier No. 8110101
  • ONLINE EXCLUSIVE PRICE
    99.00 EUR
    87.20 EUR / Pack of 1
    Save 11.80 EUR
    11% Off
 Restricted Item
Estimated Shipment: 18-01-2027
With double clamps for secure positioning.
  • Double net ruling
  • With clamps
  • Includes 2 haemocytometer glass covers
  • With LOT number on packaging
  • Labelled with: usable until, CE pursuant to IVD 98/79 EC
Specifications
Product Type Counting Chamber (Burker)
Description Double net ruling
Depth (Metric) 0.1 mm
Certifications/Compliance CE pursuant to IVD 98/79 EC
Includes 2 haemocytometer glass covers