Learn More
Marienfeld Superior™ Counting Chamber with Bright Line Double Net Ruling
Description
- Precise measuring instrument for determination of the number of particles in liquids
- Made of special optical glass
- Equipped with two grids
- With 2 haemacytometer cover glasses 20x26 mm
- Individually packed in a transparent plastic box, 10 boxes in a carton
Specifications
Specifications
| Product Type | Counting Chamber, Bright line |
| Grid Type | Neubauer |
| Description | CE-marked (IVD) |
| Cell Size | 0.0025 mm2 |
| Material | Special optical glass |
| Quantity | 1 |
| Depth (Metric) | 0.1 mm |
| Includes | Two cover glasses 20 x 26 mm |