Learn More
Invitrogen™ Countess™ Reusable Slide and Holder
Product Code. 17374933
Change view
Click to view available options
Type:
Reusable Slide
Slide Holder
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
17374933 Slide Holder Each
15311986 Reusable Slide Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17374933 Supplier Invitrogen™ Supplier No. A48208
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
The Countess Reusable Slide helps reduce costs and generate less waste when counting cells on the Countess II FL, Countess 3, and Countess 3 FL automated cell counters. The Countess 3 and 3 FL Reusable Slide Holder is required for use (also compatible with Countess II FL).
The Countess Reusable Slide offers a greener and cost-effective alternative to automated cell counting with disposable slides and is designed for use with the Countess II FL, Countess 3, and Countess 3 FL automated cell counters. The Countess 3 and 3 FL Reusable Slide Holder holds the Countess Reusable Slide and is included with the Countess 3 and 3 FL automated cell counters (also compatible with Countess II FL).
The Countess Reusable Slide is a green and cost-efficient alternative to disposable slides for automated cell counting. The reusable slide enables researchers and cell culture facilities to reduce costs and generate less waste per year, making it affordable for labs to automate their cell counts. The reusable slide is cleaned and dried between counts, just as with a hemocytometer.
Each Countess Reusable Slide has two chambers for counting one sample in duplicate or two different samples. Cell counting occurs in a central location of the counting chamber. The volume counted is 0.4 μL, the same as counting four 1-mm2 squares in a standard hemocytometer. The Countess Reusable Slide can be used with the Countess II FL, Countess 3, and Countess 3 FL automated cell counters.
The Countess 3 and 3 FL Reusable Slide Holder is included with the Countess 3 and 3 FL automated cell counters and is compatible with Countess II FL. A replacement Countess 3 and 3 FL Reusable Slide Holder (Cat. No. A48208) is available for separate purchase. Special tensioning springs hold the sample securely, ensuring a repeatable field of view (FOV) and consistent counts. The Countess 3 and 3 FL Reusable Slide Holder accepts the Countess Reusable Slide (Cat. No. A25750). It is not compatible with Countess disposable slides.
TRUSTED_SUSTAINABILITY
Specifications
Type Slide Holder
Product Line Countess
Compatibility Countess™ Reusable Slide (Cat. No. A5750)
For Use With (Equipment) Countess II FL, Countess 3 & Countess 3 FL Automated Cell Counters
Content And Storage Countess™ 3 and 3 FL Reusable Slide Holder, 1 each
Quantity 1 holder
Do you offer reusable slides and the appropriate holders for Countess II FL and Countess 3/3 FL instruments?
The Countess Reusable Slide (Cat. No. A25750) is compatible with the Countess II FL and the Countess 3/3 FL instruments. However, they require a reusable slide holder specific to the instrument. The Countess 3 & 3 FL Reusable Slide Holder (Cat. No. A48208) and the Countess II FL Reusable Slide Holder (Cat No. AMEP4746) are shipped with the instrument and can be purchased separately.
The slide holder can be held inside the back of the instrument, next to the light cube removal tool.
Where may I purchase the coverslips provided with the Countess Reusable Slide (Cat no. A25750) as a stand-alone product?
Hausser Scientific Cover Glasses/Slips (Cat. No. 02-617-581) can be purchased through Fisher Scientific; these are 20 x 26 x 0.5 mm borosilicate glass coverslips.
Can the Countess 3 & 3 FL Reusable Slide Holder (Cat no. A48208) be used on a Countess II FL Automated Cell Counter (Cat. No. AMQAF1000)?
Yes. The Countess 3 & 3 FL Reusable Slide Holder (Cat. No. A48208) be used on a Countess II FL Automated Cell Counter (Cat. No. AMQAF1000).
Can the Countess II FL Reusable Slide Holder (Cat no. AMEP4746) be used on a Countess 3 and 3 FL Automated Cell Counter (Cat. Nos. AMQAX2000 and AMQAF2000)?
Yes. The Countess II FL Reusable Slide Holder (Cat no. AMEP4746) can be used on a Countess 3 and 3 FL Automated Cell Counter (Cat. Nos. AMQAX2000 and AMQAF2000).
Can a conventional cell counting chamber be used with the Countess 3 & 3 FL Reusable Slide Holder (Cat. No. A48208)?
No. The Countess 3 & 3 FL Reusable Slide Holder (Cat. No. A48208) and the Countess instruments will only work with the Countess Reusable Slide (Cat. No. A25750). Using other slides can damage the instrument, due to differences in the height and width of the slides.