Learn More
Azpack™ Cotton Wool, Non-absorbent
Product Code. 11468153
Change view
Click to view available options
Weight (Metric):
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Weight (Metric) Unit Size
11468153 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11468153 Supplier Azpack™ Supplier No. 11468153
  • ONLINE EXCLUSIVE PRICE
    62.95 EUR
    43.24 EUR / 500g
    Save 19.71 EUR
    31% Off
Estimated Shipment: 11-12-2026
Test absorbency by placing a teaspoon of water in the hollow of a cotton. Azpack™ Cotton Wool, Non-absorbent is defatted due to bleaching.
  • 100% Bleached cotton combers
  • Treated with a non-absorbent finish
  • 1 roll is approximately 3.5m x 325mm (L x W)
  • 500g
  • CE Marked
Specifications
Product Type Nonabsorbent Cotton
Material Bleached Cotton treated with nonabsorbent finish
Grade CE Marked
Quantity 1 Ea.
Weight (Metric) 500 g