Learn More
Corning™ Costar™ Brand 96-Well EIA/RIA Plates
GREENER_CHOICE
Product Code. 10440862 Shop All Corning Products
Click to view available options
Color:
Clear
Unit Size:
Pack of 100
Product Code. 10440862
Brand: Corning™ 3361
Only null left
Add to basket
Add to basket
Flat, high binding, sterile, with lid.
  • Cell culture microplates are sterile and nonpyrogenic.
  • Lids available where indicated
Cat. nos. 3798 and 3898 processed to improve hydrophilicity for hemagglutination and similar assays.
Cat. Nos. 3585 and 3595 have special low evaporation lids.
Cat. No. 3841 is aseptically manufactured.
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Lid With Lid
Array 8 x 12
Binding Type High Bind
Color Clear
Material Polystyrene
No. of Wells 96
Volume (Metric) Working 75 to 200 μL
Sterility Sterile
Surface Treatment High Binding
Well Shape Flat Bottom (F-well)
For Use With (Application) Ideal for immunoassays
No. per Case 100/Cs.
No. per Pack 20/Pk.
Quantity 100/Cs.