missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cortisone Acetate, TRC
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Specifications
Specifications
| Chemical Name or Material | Cortisone Acetate |
| CAS | 50-04-4 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C23 H30 O6 |
| Quantity | 50 mg |
| Synonym | Cortisone, 21-acetate (8CI), Pregn-4-ene-3,11,20-trione, 17,21-dihydroxy-, 21-acetate (7CI), 17,21-Dihydroxypregn-4-ene-3,11,20-trione 21 acetate, 21-Acetoxy-17-hydroxypregn-4-ene-3,11,20-trione, 21-Acetoxy-17α-hydroxy-3,11,20-triketopregnene-4, 21-Acetoxy-17α-hydroxypregn-4-ene-3,11,20-trione, 4-Pregnene-17a,21-diol-3,11,20-trione 21-acetate, Adreson, Artriona, Biocort Acetate, Compound E Acetate, Corlin, Cortadren, Cortelan, Cortisone acetate, Cortistab, Cortisyl, Cortisyl Artriona, Cortone acetate, Incortin, Irisone acetate, NSC 49420, Ricortex, Scheroson |
| InChI Formula | InChI=1S/C23H30O6/c1-13(24)29-12-19(27)23(28)9-7-17-16-5-4-14-10-15(25)6-8-21(14,2)20(16)18(26)11-22(17,23)3/h10,16-17,20,28H,4-9,11-12H2,1-3H3/t16-,17-,20+,21-,22-,23-/m0/s1 |
| SMILES | CC(=O)OCC(=O)[C@@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3C(=O)C[C@]12C |
| IUPAC Name | [2-[(8S,9S,10R,13S,14S,17R)-17-hydroxy-10,13-dimethyl-3,11-dioxo-1,2,6,7,8,9,12,14,15,16-decahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] acetate |
| Molecular Weight (g/mol) | 402.48 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction