missing translation for 'onlineSavingsMsg'
Learn More

Cortisone Acetate, TRC

Product Code. 30365100
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy

Product Code. 30365100

Brand: Toronto Research Chemicals TRCC6965101G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cortisone Acetate
CAS 50-04-4
Purity Grade Notes HPLC
Molecular Formula C23 H30 O6
Quantity 1 g
Synonym Cortisone, 21-acetate (8CI), Pregn-4-ene-3,11,20-trione, 17,21-dihydroxy-, 21-acetate (7CI), 17,21-Dihydroxypregn-4-ene-3,11,20-trione 21 acetate, 21-Acetoxy-17-hydroxypregn-4-ene-3,11,20-trione, 21-Acetoxy-17α-hydroxy-3,11,20-triketopregnene-4, 21-Acetoxy-17α-hydroxypregn-4-ene-3,11,20-trione, 4-Pregnene-17a,21-diol-3,11,20-trione 21-acetate, Adreson, Artriona, Biocort Acetate, Compound E Acetate, Corlin, Cortadren, Cortelan, Cortisone acetate, Cortistab, Cortisyl, Cortisyl Artriona, Cortone acetate, Incortin, Irisone acetate, NSC 49420, Ricortex, Scheroson
InChI Formula InChI=1S/C23H30O6/c1-13(24)29-12-19(27)23(28)9-7-17-16-5-4-14-10-15(25)6-8-21(14,2)20(16)18(26)11-22(17,23)3/h10,16-17,20,28H,4-9,11-12H2,1-3H3/t16-,17-,20+,21-,22-,23-/m0/s1
SMILES CC(=O)OCC(=O)[C@@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3C(=O)C[C@]12C
IUPAC Name [2-[(8S,9S,10R,13S,14S,17R)-17-hydroxy-10,13-dimethyl-3,11-dioxo-1,2,6,7,8,9,12,14,15,16-decahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] acetate
Molecular Weight (g/mol) 402.48
Formula Weight 402.2042
Percent Purity >95
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.