Learn More
Corning™ Sterile Microspatulas, rounded end
BIOPHARMAGREENER_CHOICE
Product Code. 10486572 Shop All Corning Products
Change view
Click to view available options
Length (Metric):
15.2 cm
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
10486572 15.2 cm Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10486572 Supplier Corning™ Supplier No. 3013
  • ONLINE EXCLUSIVE PRICE
    169.00 EUR
    157.21 EUR / Pack of 50
    Save 11.79 EUR
    6% Off
View all promotions
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
Constructed of rigid nontoxic polystyrene to provide good chemical resistance to aqueous solutions. Corning™ Sterile Microspatulas are an ideal solution for molecular biology applications, lyophilized samples, and contamination-free sampling.
  • Corning spatulas are designed to save time and to provide contamination-free samples.
  • Eliminates the recycling and resterilizing necessary with reusable spatulas.
  • Available in five different configurations.
  • Microspatulas available in two configurations.
  • Individually packaged.
  • Antistatic.
  • Sterile.
  • RNase-/DNase-free.
  • Nonpyrogenic.
TRUSTED_SUSTAINABILITY
Specifications
Sterility Sterile
Certifications/Compliance USP Class VI, non-pyrogenic, RNase and DNase-free Certified
Disposable Single-use
Product Type Micro Spatula
For Use With (Application) Molecular biology applications, lyophilized samples and contamination-free sampling
Length (English) 5.98 in.
Includes V-shaped scoop at one end and a flat rounded blade at the other end
Material Polystyrene
Length (Metric) 15.2 cm