Includes
Pipet tips in rack
- IsoTip filtered pipet tips feature an inert, hydrophobic barrier that prevents aerosolized contaminants from coming in contact with pipettor shafts. (A Pipet Tip Compatibility Guide can be requested or downloaded from the Corning website.)
- Ideal for applications where avoiding cross contamination is critical, such as DNA amplification and radioisotope handling.
- Packaged sterile.
- RNase-/DNase-free.
- Nonpyrogenic.
Specifications
Specifications
| Volume (Metric) | 0.1 to 2 μL |
| Volume (Metric) Dispensing | 0.1 to 2 μL |
| Filtered | Filtered |
| Sterility | Sterile |
| Color | Natural |
| Autoclavable | Not Autoclavable |
| Certifications/Compliance | ISO 9001 |
| Clean Claims | RNase-/DNase-free, Nonpyrogenic |
| For Use With (Application) | DNA Amplification, Radioisotope Handling |
| Format | Rack |
| Show More |