Learn More
Corning™ Filtered IsoTip™ Universal Fit Racked Pipet Tips
Product Code. 10173711 Shop All Corning Products
Change view
Click to view available options
Volume (Metric):
0.1 to 2 μL
0.2 to 10 μL
0.5 to 10 μL
1 to 200 μL
1 to 30 μL
100 to 1000 μL
Unit Size:
Pack of 1000
Pack of 960
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
10173711 0.5 to 10 μL Pack of 960
10674713 0.1 to 2 μL Pack of 960
10452181 1 to 30 μL Pack of 960
10035510 0.2 to 10 μL Pack of 960
10116840 100 to 1000 μL Pack of 1000
10106840 1 to 200 μL Pack of 960
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10173711 Supplier Corning™ Supplier No. 4808
Add to basket
Add to basket
Includes
Pipet tips in rack
Ideal for applications where avoiding cross-contamination is critical, such as DNA amplification and radioisotope handling
  • IsoTip filtered pipet tips feature an inert, hydrophobic barrier that prevents aerosolized contaminants from coming in contact with pipettor shafts. (A Pipet Tip Compatibility Guide can be requested or downloaded from the Corning website.)
  • Ideal for applications where avoiding cross contamination is critical, such as DNA amplification and radioisotope handling.
  • Packaged sterile.
  • RNase-/DNase-free.
  • Nonpyrogenic.
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 0.5 to 10 μL
Volume (Metric) Dispensing 0.5 to 10 μL
Filtered Filtered
Sterility Sterile
Color Natural
Autoclavable Not Autoclavable
Certifications/Compliance ISO 9001
Clean Claims RNase-/DNase-free, Nonpyrogenic
For Use With (Application) DNA Amplification, Radioisotope Handling
Format Rack
Tip Style Round
Packaging Rack
Quantity 96/Pk.
Product Type Filtered Pipette Tips
Includes 10 Racks x 96 Tips
Green Features Facilities 100% Covered By Renewable Energy
Disposable Single-use
For Use With (Equipment) Eppendorf, Universal Fit Ultramicro
Graduated Yes
Product Line Isotip
Material Polypropylene
Shape Round