Description
- Polycarbonate and nonsterile
- O-ring and retaining clips assure the reusable chamber remains watertight in waterbaths and airtight in hybridization ovens
- Wells in the base hold 10μL of water to maintain optimal interior humidity
- Increased interior depth allows for single slide hybridizations and for user to place two arrays face-to-face and hybridize using a single labeled target
- Used with raised-edge coverslips that are thicker or taller than standard thin coverglass
Specifications
Specifications
| Quantity | 5 Case |
| Material | Polycarbonate |
| Type | Hybridization Chamber |
| For Use With (Application) | Wells in the base hold 10μL of water to maintain optimal interior humidity, increased interior depth allows for single slide hybridizations and for user to place two arrays face-to-face and hybridize using a single labeled target |