Learn More
Corning™ Frosted Microscope Slides
Product Code. 11363835 Shop All Corning Products
Change view
Click to view available options
Frosted:
One End, Both Sides
One end, one side
Unit Size:
Pack of 1440
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Frosted Unit Size
11363835 One End, Both Sides Pack of 1440
11343835 One end, one side Pack of 1440
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11363835 Supplier Corning™ Supplier No. 294975X25
  • ONLINE EXCLUSIVE PRICE
    710.00 EUR
    662.37 EUR / Pack of 1440
    Save 47.63 EUR
    6% Off
Estimated Shipment: 01-10-2026
Made from water-white glass to maximize clarity. Easy to write on.
These Corning™ microscope slides are made from water-white glass to maximize clarity and are frosted at one end, both sides. Slides are 0.9 to 1.10 mm thick. Corning™ frosted slides are highly legible and easy to write on because they are sandblasted instead of etched. To minimize waste, they are inspected for chips and rough edges before being packaged. Products are packaged in 1/2 gross boxes (approx. 72 slides each). 10 gross = 20 boxes (1440 slides total)
TRUSTED_SUSTAINABILITY
Specifications
Length (Metric) 75 mm
Width (Metric) 25 mm
No. per Case 1440
No. per Pack 72
Frosted One End, Both Sides
Material Soda Lime Glass
Thickness (Metric) 0.9 to 1.1 mm