Learn More
Corning™ Flexible Packaging Systems: MPC Connector Tubing
Product Code. 17243594 Shop All Corning Products
Change view
Click to view available options
Tubing Style:
Fits tubing 60.96 cm
Fits tubing 91.44 cm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Tubing Style unitSize
17243594 Fits tubing 91.44 cm Each
17253594 Fits tubing 60.96 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17243594 Supplier Corning™ Supplier No. 9170004
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Clear tubing; female MPC connector
  • Chemically resistant
  • Heat sealable
  • Flexible
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Male luer Cap
Tubing Style Fits tubing 91.44 cm
Product Type Connector tubing
Certifications/Compliance cGMPs, FDA Regulation 21 CFR part 820 and ISO 13485 Quality
Color Clear