Learn More
Corning™ Costar™ Thermowell™ Plate Holder
Product Code. 10206631 Shop All Corning Products
Change view
Click to view available options
For Use With (Equipment):
Thermo well polycarbonate plates
Unit Size:
Pack of 20
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
10206631 Thermo well polycarbonate plates Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10206631 Supplier Corning™ Supplier No. 2572
  • ONLINE EXCLUSIVE PRICE
    94.25 EUR
    88.87 EUR / Pack of 20
    Save 5.38 EUR
    5% Off
Estimated Shipment: 17-02-2027
For Thermo well polycarbonate plates
  • 96-well egg crate style holder for 1 x 8 Stripwell™ strips
  • Holds twelve 1 x 8 strips
  • Allows each individual well to be positioned back into the plate once broken apart
TRUSTED_SUSTAINABILITY
Specifications
Type Plate Holder
For Use With (Equipment) Thermo well polycarbonate plates