Learn More
Corning™ Color-Coded Cap Inserts
GREENER_CHOICE
Product Code. 10461822 Shop All Corning Products
Change view
Click to view available options
Closure Color:
Assorted
Blue
Green
Red
White
Yellow
Unit Size:
Pack of 500
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Closure Color unitSize
10461822 Assorted Pack of 500
10474912 Blue Pack of 500
10317532 White Pack of 500
10664933 Yellow Pack of 500
10032202 Green Pack of 500
10377572 Red Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10461822 Supplier Corning™ Supplier No. 430499
Currently Unavailable: We have no information when this product will be available.
View alternative products
For use with Corning cryogenic vials except 03-374-6
  • Cap inserts provide color coding for easy sample identification.
  • Inserts are packaged in resealable bags.
  • Nonsterile
  • Cap inserts fit all Corning™ cryogenic vials.
TRUSTED_SUSTAINABILITY
Specifications
Closure Color Assorted
Color Assorted
Material Polypropylene
Sterility Non-sterile
No. per Case 500
No. per Pack 50
Closure Material Polypropylene