Learn More
Corning™ Collagen IV, Human
Product Code. 11523560 Shop All Corning Products
Change view
Click to view available options
Quantity:
0.25 mg
Unit Size:
250µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11523560 0.25 mg 250µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11523560 Supplier Corning™ Supplier No. 354245
  • ONLINE EXCLUSIVE PRICE
    597.00 EUR
    566.23 EUR / 250µg
    Save 30.77 EUR
    5% Off
 Restricted Item
In Stock
Used as either a thin layer on tissue-culture surfaces to enhance cell attachment and proliferation, or as a gel to promote expression of cell-specific morphology and function
Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.
Corning offers a broad range of collagen types that are derived from multiple species. These collagens have been used to culture a variety of cell types to promote adhesion, growth, and/or differentiation.
TRUSTED_SUSTAINABILITY
Specifications
Concentration 1 to 5 μg/cm2
For Use With (Application) Cell Attachment and Proliferation
Formulation Frozen in 100 mm borate buffer pH 8.0
pH Range 8
Quantity 0.25 mg
Source Human Placenta
Product Type Collagen, Human Type IV
Content And Storage -70°C