Learn More
Corning™ Cell Culture Buffers: Dulbecco's Phosphate-Buffered Salt Solution 1X
Description
Dulbecco's Phosphate Buffered Saline (DPBS) is a buffer solution which can be used to maintain cell culture media in the physiological pH range of 7.0 to 7.6. It can serve as an irrigating, transporting or diluting fluid while maintaining cell tonicity and viability for a limited period of time.
Specifications
Specifications
| Chemical Name or Material | DPBS |
| Concentration | 1 X |
| pH | 7.3 to 7.5 |
| Physical Form | Liquid |
| Quantity | 6 x 1000 mL |
| Solution Type | Buffer |
| Formulation | Potassium Chloride 0.2g/L, Potassium Dihydrogen Phosphate 0.2g/L, Sodium Chloride 8g/L, Disodium Phosphate 1.15g/L |
| Includes | No Calcium, No Magnesium |
| Grade | Not Available |