Learn More
Corning™ 96-Well Round Bottom Polystyrene Ulta-Low Attachment Microplate
Product Code. 17750708 Shop All Corning Products
Change view
Click to view available options
Color:
Black
Unit Size:
Pack of 24
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17750708 Black Pack of 24
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17750708 Supplier Corning™ Supplier No. 4591
  • ONLINE EXCLUSIVE PRICE
    695.00 EUR
    646.84 EUR / Pack of 24
    Save 48.16 EUR
    6% Off
Estimated Shipment: 28-10-2026
Ulta-Low attachment surface with covalently bonded hyrogel minimizes cell attachment, protein absorption, enzyme activation and cellular activation
  • Designed to reduce well-to-well cross-talk
  • White microplates enhance luminescent signals and have low background luminescence.
  • Black microplates have low background fluorescence and minimize light scattering.
  • TRUSTED_SUSTAINABILITY
    Specifications
    Format Standard
    Lid With Lid
    Array 8 x 12
    Color Black
    Material Polystyrene
    Volume (Metric) Well 360 μL
    No. of Wells 96
    Volume (Metric) Working 75 to 200 μL
    Sterility Sterile
    Well Shape Round
    No. per Case 24/Cs.
    No. per Pack 1/Pk.