Learn More
Fisherbrand™ Cork Rings
Product Code. 12376447 Shop All Fisherbrand Products
Change view
Click to view available options
Diameter (Metric) Inner:
45 mm
85 mm
120 mm
160 mm
200 mm
Diameter (Metric) Outer:
75 mm
115 mm
150 mm
195 mm
240 mm
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner Diameter (Metric) Outer unitSize
12376447 160 mm 195 mm Each
12366447 120 mm 150 mm Each
12346447 45 mm 75 mm Each
12386447 200 mm 240 mm Each
12356447 85 mm 115 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 12376447
  • ONLINE EXCLUSIVE PRICE
    21.00 EUR
    13.92 EUR / Pack of 1
    Save 7.08 EUR
    33% Off
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
Ring to support flasks
Ring to support flasks
Available in different diameters
Specifications
For Use With (Application) Flask Support
Quantity 1 Each
Depth (Metric) 38 mm
Diameter (Metric) Outer 195 mm
Product Type Cork Ring
Diameter (Metric) Inner 160 mm