Learn More
Brady™ Core Paper Labels, 76 mm
Product Code. 31751190
Change view
Click to view available options
Dimensions (L x W):
101.60 x 101.60 mm
203.20 x 101.60 mm
Unit Size:
1 roll
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) unitSize
31751190 101.60 x 101.60 mm 1 roll
31751199 203.20 x 101.60 mm 1 roll
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 31751190 Supplier Brady™ Supplier No. 324910
Add to basket
Add to basket
Printable, paper labels for direct thermal printers
  • Ideal label sizing for logistical applications.
Specifications
Adhesive Type Permanent Acrylic
Coating None
Color White
Finish Matte
Format Die-cut
Material Paper
Printable Sides Single-side Printable
Print Technology Direct Thermal
Product Type Label
Sterility Non-sterile
For Use With (Equipment) Brady i6100
No. per Pack 3000
Shape Rectangle
Thickness (English) 0.0039 in.
Thickness (Metric) 0.099 mm
Dimensions (L x W) 101.60 x 101.60 mm
Length (English) 101.60 mm
Width (English) 4 in.
Width (Metric) 101.60 mm
Width (English) Printable Area 101.60 mm
Height (English) 4 in.
Height (Metric) 101.60 mm
Alternate Part Number i61-DT-77-7604-1.5