missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated CPT1B-specific Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
129.00 EUR - 450.00 EUR
Specifications
| Antigen | CPT1B-specific |
|---|---|
| Concentration | 1000 μg/mL |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | CoraLite Plus 488 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30236313
|
Proteintech
CL488-22170-20UL |
20 μL |
140.00 EUR 129.00 EUR / 20µL Save 11.00 EUR 7% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30237063
|
Proteintech
CL488-22170-100UL |
100 μL |
450.00 EUR
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The CPT1B-specific antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CPT1B-specific. This antibody recognizes human, mouse, rat antigen. The CPT1B-specific antibody has been validated for the following applications: IF-P analysis. This antibody is coralite™ plus 488.Specifications
| CPT1B-specific | |
| Immunofluorescence | |
| CoraLite Plus 488 | |
| Rabbit | |
| Human, Mouse, Rat | |
| CPT1-M; FLJ55729; FLJ58750; KIAA1670; M-CPT1; MCCPT1 | |
| IgG | |
| Antigen affinity purification | |
| Conjugated Antibody |
| 1000 μg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| 1375 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title