missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated CPT1B-specific Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-22170-100UL
This item is not returnable.
View return policy
Description
The CPT1B-specific antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CPT1B-specific. This antibody recognizes human, mouse, rat antigen. The CPT1B-specific antibody has been validated for the following applications: IF-P analysis. This antibody is coralite™ plus 488.Specifications
| CPT1B-specific | |
| Polyclonal | |
| CoraLite Plus 488 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 1375 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Conjugated Antibody | |
| IgG |
| Immunofluorescence | |
| 1000 μg/mL | |
| CPT1-M; FLJ55729; FLJ58750; KIAA1670; M-CPT1; MCCPT1 | |
| Recombinant Protein | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| 75-85 kDa | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction