missing translation for 'onlineSavingsMsg'
Learn More

Coptisine chloride, MedChemExpress

Product Code. 16469078
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16469078 10 mg 10mg
16464877 50 mg 50mg
16409136 100 mg 100mg
16401498 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16469078 Supplier Medchem Express Supplier No. HYN0736

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Coptisine chloride is an alkaloid from Chinese goldthread, and acts as an efficient uncompetitive IDO inhibitor with a Ki value of 5.8 μM and an IC50 value of 6.3 μM. Coptisine chloride is a potent H1N1 neuraminidase (NA-1) inhibitor with an IC50 of 104.6 μg/mL and can be used for influenza A (H1N1) infection.

Coptisine chloride is an alkaloid from Chinese goldthread, and acts as an efficient uncompetitive IDO inhibitor with a Ki value of 5.8 μM and an IC50 value of 6.3 μM. Coptisine chloride is a potent H1N1 neuraminidase (NA-1) inhibitor with an IC50 of 104.6 μg/mL and can be used for influenza A (H1N1) infection.

Specifications

Chemical Name or Material Coptisine chloride
CAS 6020-18-4
Color Reddish Brown
Purity Grade Notes Research
Molecular Formula C19H14ClNO4
Quantity 10 mg
Solubility Information DMSO : 10.42 mg/mL (29.29 mM; ultrasonic and warming and heat to 60°C)
SMILES C1(C(CC[N+]2=C1C=C(C=C3)C(C4=C3OCO4)=C2)=C5)=CC6=C5OCO6.[Cl-]
Molecular Weight (g/mol) 355.77
Formula Weight 355.77
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.