missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) Sulfate, Pentahydrate, ACS, 99.0 to 100.5%, MilliporeSigma™

Product Code. 15867130
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
50 kg
Unit Size:
1kg
250g
50kg
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15867130 50 kg 50kg
15834300 250 g 250g
15844300 1 kg 1kg
15854300 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15867130 Supplier Supelco Supplier No. 1027909050
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7758-99-8 | CuH10O9S | 249.68 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-99-8
Molecular Formula CuH10O9S
Molecular Weight (g/mol) 249.68
MDL Number MFCD00149681
InChI Key JZCCFEFSEZPSOG-UHFFFAOYSA-L
Synonym copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas
PubChem CID 24463
ChEBI CHEBI:31440
IUPAC Name copper(2+) pentahydrate sulfate
SMILES O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O

Specifications

Color Blue
Physical Form Crystals
Quantity 50 kg
Assay Percent Range 99.0 to 100.5%
Linear Formula CuSO4 · 5H2O
Formula Weight 249.68
Percent Purity 99.0 to 100.5%
Grade ACS/EP/ISO/Reagent
Packaging Plastic Bottle
Density 2.284g/cm3 (20°C)
Chemical Name or Material Copper(II) Sulfate, Pentahydrate, EMSURE™, For Analysis
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.