missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) sulfate pentahydrate, ACS, 98.0-102.0%

Product Code. p-3256990
Change view
Click to view available options
Quantity:
500 g
2 kg
Unit Size:
2kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11367767 500 g 500g
11377767 2 kg 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11367767 Supplier Thermo Scientific Alfa Aesar Supplier No. 014178.36
Add to basket
This item is not returnable. View return policy

CAS: 7758-99-8 | CuH10O9S | 249.68 g/mol

Copper(II) sulfate, pentahydrate can be added to potassium permanganate to give an oxidant for the conversion of primary alcohols. It is used to etch zinc or copper plates for intaglio printmaking and as a mordant in vegetable dyeing. In analytical chemistry, it is used in Fehling's solution and Benedict's solution to test for reducing sugars. Also, it is used to test protein in Biuret test.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-99-8
Molecular Formula CuH10O9S
Molecular Weight (g/mol) 249.68
MDL Number MFCD00149681
InChI Key JZCCFEFSEZPSOG-UHFFFAOYSA-L
Synonym copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas
PubChem CID 24463
ChEBI CHEBI:31440
IUPAC Name copper;sulfate;pentahydrate
SMILES O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O

Specifications

Physical Form Crystalline
Quantity 500 g
Assay Percent Range 98.0-102.0%
Solubility Information Soluble in water,methanol and glycerol.
Formula Weight 249.68 (159.60 Anhydrous)
Grade ACS
Density 2.27
Chemical Name or Material Copper(II) sulfate, pentahydrate
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Copper(II) sulfate pentahydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Copper(II) sulfate pentahydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.