missing translation for 'onlineSavingsMsg'
Learn More

Copper(Ii) Sulfate Hydrate, 98%, Honeywell Fluka™

Product Code. p-7120355 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Poly Bottle
Unit Size:
1kg
5kg
This item is not returnable. View return policy

Product Code. 15601900

Brand: Honeywell-Fluka 2092011KG

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

>98%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23254-43-5
Molecular Formula CuO4S
Molecular Weight (g/mol) 159.60
MDL Number MFCD00149682
InChI Key ARUVKPQLZAKDPS-UHFFFAOYSA-L
Synonym copper sulfate monohydrate, cupric sulfate monohydrate, copper sulfate, monohydrate, copper sulfate hydrate, caswell no. 257, epa pesticide chemical code 024402, copper ii sulfate hydrate, sulfuric acid copper 2+ salt 1:1 , monohydrate, copper sulphate hydrate, cuso4.h2o
PubChem CID 61499
IUPAC Name copper;sulfate;hydrate
SMILES [Cu++].[O-]S([O-])(=O)=O

Specifications

Melting Point 3603g/cu.cm
Color Blue or White
Quantity 1 kg
Linear Formula CuSO4 · xH2O
UN Number UN3077
Formula Weight 159.61g/mol (anhydrous basis)
Percent Purity 98%
Packaging Poly Bottle
Appearance Crystals/Powder
Vapor Pressure 7.3mmHg (25°C)
Density 200°C
Chemical Name or Material Copper(Ii) Sulfate Hydrate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.