accessibility menu, dialog, popup

UserName

CAS RN 23254-43-5

OSOOOCu

CAS RN 23254-43-5

IUPAC Name: copper(2+) hydrate sulfate
Synonyms: copper(2+) hydrate sulfate
Molecular Weight (g/mol): 177.62
Molecular Formula: CuH2O5S
InChi Key: CYKLGTUKGYURDP-UHFFFAOYSA-L
SMILES: O.[Cu++].[O-]S([O-])(=O)=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)