Learn More
Copper(I) Cyanide, Sigma-Aldrich™
Product Code. 15862351
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15862351 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
MQ200 Graded - For Research and Non-regulated Industrial Applications
Product Code. 15862351 Supplier Sigma Aldrich Supplier No. 8418110100
 Restricted Item
Estimated Shipment: 25-11-2026
MQ200 Graded - For Research and Non-regulated Industrial Applications
For synthesis
TRUSTED_SUSTAINABILITY
Specifications
Melting Point 473°C
CAS 544-92-3
Quantity 100 g
Density 2.92 g/cm3
MDL Number MFCD00010975
Synonym Copper(I) cyanide, Cupricin, Copper(I) cyanide
Molecular Weight (g/mol) 89.56
Grade Research
Chemical Name or Material Copper(I) Cyanide