Learn More
Copper flake, -325 mesh, 99% (metals basis)
Product Code. 15432137
Change view
Click to view available options
Quantity:
250 g
500 g
1 kg
Unit Size:
1kg
250g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15432137 1 kg 1kg
15412137 250 g 250g
15422137 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15432137 Supplier Thermo Scientific Alfa Aesar Supplier No. 045073.A1
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
CAS: 7440-50-8 | Cu | 63.55 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7440-50-8
Molecular Formula Cu
Molecular Weight (g/mol) 63.55
MDL Number MFCD00010965
InChI Key RYGMFSIKBFXOCR-UHFFFAOYSA-N
Synonym cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
PubChem CID 23978
ChEBI CHEBI:30052
IUPAC Name copper
SMILES [Cu]
Specifications
Melting Point 1083°C
Boiling Point 2562°C
Appearance Copper colored flakes
Density 8.94 g/cm3
Quantity 1 kg
Mesh Size -325 Mesh
Odor Odorless
Vapor Pressure 0 hPa
Assay Percent Range 99% (Metals basis)
Form Flake
Formula Weight 63.55
Chemical Name or Material Copper