Learn More
Copper Aluminum Oxide (Technical Grade), TRC
Product Code. 30367627
Change view
Click to view available options
Quantity:
2.5 g
5 g
25 g
Unit Size:
2.5g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30367627 25 g -
30355238 2.5 g 2.5g
30361608 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30367627 Supplier Toronto Research Chemicals Supplier No. TRCC99030825G
 Restricted Item
Estimated Shipment: 24-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemische Identifikatoren
CAS 12042-92-1
Molecular Formula 2 Al . Cu . 2 H2 O . 2 O
Molecular Weight (g/mol) 185.538
Synonym Copper(II) Aluminate
IUPAC Name copper;alumane;oxygen(1-);dihydrate
SMILES O.O.[Cu+2].[O-].[O-].[Al].[Al]
Spezifikation
Quantity 25 g
InChI Formula InChI=1S/2Al.Cu.2H2O.2O.6H/h;;;2*1H2;;;;;;;;/q;;+2;;;2*-1;;;;;;
Formula Weight 184.904
Chemical Name or Material Copper Aluminum Oxide (Technical Grade)