Learn More
Corning™ CoolCell™ LX Cell Freezing Vial Containers
BIOPHARMA
Product Code. 15552771
Change view
Click to view available options
Color:
Assorted (Green, Orange, Pink, Purple)
Green
Orange
Pink
Purple
Quantity:
1/Pk., 1/Cs.
4/Pk., 1/Cs.
Unit Size:
Each
Pack of 4
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Quantity Unit Size
15552771 Purple 1/Pk., 1/Cs. Each
15562771 Green 1/Pk., 1/Cs. Each
15572771 Orange 1/Pk., 1/Cs. Each
15582771 Pink 1/Pk., 1/Cs. Each
15813885 Assorted (Green, Orange, Pink, Purple) 4/Pk., 1/Cs. Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 15552771 Supplier Corning™ Supplier No. 432001
  • ONLINE EXCLUSIVE PRICE
    219.00 EUR
    206.20 EUR / Pack of 1
    Save 12.80 EUR
    5% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket
Provides alcohol-free freezing at the rate of -1°C/minute when placed in a -80°C freezer.
Corning CoolCell, in combination with a -80°C freezer, will provide alcohol-free freezing at the rate of -1°C/minute that is ideal for cryopreservation of most cells and cell lines. Using a combination of uniform-density cross-linked polyethylene foam, a solid state core, and radial vial symmetry, freezing profiles are consistent and reproducible. The foam is non-absorbent and will impose negligible change in the freezer environment; thereby protecting nearby frozen samples. The removal of vials is accomplished by tipping the device upside down so the vials slide out.
Features: Unique features of controlled-rate freezing with Corning™ CoolCell™ include:
  • Ease of use
  • Alcohol and fluid-free freezing
  • Lower cost of use than alcohol-based devices
  • High cell recovery and cell viability
  • Reproducibility
  • Simple, consistent way to standardize controlled-rate freezing
TRUSTED_SUSTAINABILITY
Specifications
Color Purple
Material Polyethylene Foam
Shape Round
Type Freezer Box
For Use With (Application) Cryopreservation of Most Cells and Cell Lines
Holds 12 (Standard 1 to 2 mL Cryogenic Vials)
Quantity 1/Pk., 1/Cs.
Dimensions (L x W) 11.7 x 10.11 cm
Length (Metric) 11.7 cm
Width (Metric) 10.11 cm
Diameter (Metric) Well 12.7 mm
Weight (Metric) 0.19 kg