Learn More
Lonza™ Control Standard Endotoxin
Product Code. 11610241
Change view
Click to view available options
Quantity:
25 vials
Unit Size:
Pack of 25
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11610241 25 vials Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11610241 Supplier Lonza™ Supplier No. E50643L
  • ONLINE EXCLUSIVE PRICE
    2950.00 EUR
    2565.23 EUR / Pack of 25
    Save 384.77 EUR
    13% Off
 Restricted Item
Estimated Shipment: 07-12-2026
For Kinetic-QCL™ Bulk Kinetic Chromogenic LAL, E. coli Strain O55:B5.
  • Used in combination with Kinetic-QCL™ Bulk Kinetic Chromogenic LAL Assay, K50-643L and K50-643U.
  • It is available as a made-to-order product.
TRUSTED_SUSTAINABILITY
Specifications
Color White
Description Powder
Cell Type E. Coli strain O55:B5.
For Use With (Application) Bulk Kinetic Chromogenic LAL Assay, K50-643L and K50-643U
Quantity 25 vials