Learn More
Buerkle™ Close-it™ Control Seal Food & Pharma
Product Code. 15321306 Shop All Bürkle Products
Change view
Click to view available options
Color:
Black
Blue
Green
Unit Size:
Pack of 500
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
15321306 Blue Pack of 500
15346287 Green Pack of 500
15356287 Black Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15321306 Supplier Buerkle™ Supplier No. 53032021
Estimated Shipment: 27-11-2026
Control Seal Close-it Food and Pharma with an adhesive approved for use with foodstuffs. Reliably prevents products from leaking out after sampling.
  • Reliably prevents products from leaking out after sampling.
  • The adhesive complies with the European guidelines for foodstuffs.
  • Aluminum foil barrier layer
  • Suitable for sampling foodstuffs, pharmaceuticals, cosmetics, food additives, even frozen goods – in short, for all applications in which cleanliness is particularly important.
  • According to FDA Guidelines
  • Particularly suited for the food and pharma industry
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance FDA
Color Blue
Product Type Control Seal
For Use With (Application) Foodstuffs
Shape Square
Dimensions (L x W) 95 x 95 mm
Weight (Metric) 0.8965 kg